Thermo Scientific Chemicals

μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals

Catalog number: 197591000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals

Catalog number: 197591000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 197591000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12081-16-2
IUPAC Name
bis(chlororhodium); tetrakis(ethene)
Molecular Formula
C8H16Cl2Rh2
InChI Key
OOQJCPOXJFCGCR-UHFFFAOYSA-L
SMILES
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Appearance (Color)
Orange to rust brown to black
Rhodium (Rh)
>=52.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text