Thermo Scientific Chemicals

Tetracarbonyldi-μ-chlorodirhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 197600010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetracarbonyldi-μ-chlorodirhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 197600010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197600010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14523-22-9
IUPAC Name
bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
Molecular Formula
C4Cl2O4Rh2
InChI Key
FGKDXBICTVUSPK-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Appearance (Color)
Red to brown
Assay
>=96.0 % (ex Rh)
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals or needles

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text