Thermo Scientific Chemicals

Potassium hexachloroosmate(IV), min 39% Os, Thermo Scientific Chemicals

Catalog number: 197650050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium hexachloroosmate(IV), min 39% Os, Thermo Scientific Chemicals

Catalog number: 197650050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197650050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16871-60-6
IUPAC Name
osmium(4+) dipotassium hexachloride
Molecular Formula
Cl6K2Os
InChI Key
VGKQJCSDERXWRV-UHFFFAOYSA-H
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Os+4]
Appearance (Color)
Bordeaux to purple
Appearance (Form)
Powder
Assay
>=39 % (Os)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text