Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 197680025
Price (USD)/ Each
3,439.65
Online exclusive
3822.00 
Save 382.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
3,439.65
Online exclusive
3822.00 
Save 382.35 (10%)
Add to cart
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals
Catalog number197680025
Price (USD)/ Each
3,439.65
Online exclusive
3822.00 
Save 382.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.1 % (1 g, 70°C, 2 h)
Chloride (Cl)=<5 ppm
Calcium (Ca)=<10 ppm
Cadmium (Cd)=<1 ppm
Chromium (Cr)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only