Copper(II) sulfate, 98+%, for analysis, anhydrous
Copper(II) sulfate, 98+%, for analysis, anhydrous
Copper(II) sulfate, 98+%, for analysis, anhydrous
Copper(II) sulfate, 98+%, for analysis, anhydrous
Thermo Scientific Chemicals

Copper(II) sulfate, 98+%, for analysis, anhydrous

CAS: 7758-98-7 | CuO4S | 159.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 197711000
Price (USD)
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
71.20
Save 13.55 (19%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
71.20
Save 13.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<50 ppm
Loss on drying=<1 % (300°C)
Total nitrogen (as N)=<0.01 %
Calcium (Ca)=<20 ppm
Magnesium (Mg)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Share catalog number, name or link