Copper(II) sulfate pentahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Copper(II) sulfate pentahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) sulfate pentahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 197720050
Price (USD)/ Each
335.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
395.00 
Save 59.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
335.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
395.00 
Save 59.35 (15%)
Add to cart
Copper(II) sulfate pentahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number197720050
Price (USD)/ Each
335.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
395.00 
Save 59.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecopper(2+) pentahydrate sulfate
Molecular FormulaCuH10O9S
InChI KeyJZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)249.68
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Chloride (Cl)=<10 ppm
Nickel (Ni)=<20 ppm
Iron (Fe)=<50 ppm
Total nitrogen (as N)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only