1x1 image pixel for data collection
Lithium carbonate, 99%, pure
Lithium carbonate, 99%, pure
Lithium carbonate, 99%, pure
Lithium carbonate, 99%, pure
Thermo Scientific Chemicals

Lithium carbonate, 99%, pure

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 197780025
Price (USD)
555.65
Special offer
Online exclusive
Ends: 26-Sep-2025
686.00
Save 130.35 (19%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
555.65
Special offer
Online exclusive
Ends: 26-Sep-2025
686.00
Save 130.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS554-13-2
IUPAC Namedilithium(1+) carbonate
Molecular FormulaCLi2O3
InChI KeyXGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Loss on ignition=<0.7 % (500°C, 30 min)
Titration excess HCl>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0445131Certificate of AnalysisMay 17, 2025197785000, 197780100, 197780025
A0396909Certificate of AnalysisDec 14, 2022197785000, 197780100, 197780025
A0403730Certificate of AnalysisDec 13, 2022197785000, 197780100, 197780025
A0413802Certificate of AnalysisDec 13, 2022197785000, 197780100, 197780025
A0410361Certificate of AnalysisDec 13, 2022197785000, 197780100, 197780025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link