Thermo Scientific Chemicals

Lithium carbonate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 197780100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Lithium carbonate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 197780100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 197780100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-13-2
IUPAC Name
dilithium(1+) carbonate
Molecular Formula
CLi2O3
InChI Key
XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]C([O-])=O
Appearance (Form)
Crystalline powder
Titration excess HCl
>=98.5 %
Appearance (Color)
White
Loss on ignition
=<0.7 % (500°C, 30 min)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text