Thermo Scientific Chemicals

2-Fluorobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 197960250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Fluorobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 197960250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197960250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-48-0
IUPAC Name
1-(bromomethyl)-2-fluorobenzene
Molecular Formula
C7H6BrF
InChI Key
FFWQLZFIMNTUCZ-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1CBr
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5495 to 1.5515 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text