Thermo Scientific Chemicals

4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 197970050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 197970050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 197970050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
459-46-1
IUPAC Name
1-(bromomethyl)-4-fluorobenzene
Molecular Formula
C7H6BrF
InChI Key
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
SMILES
FC1=CC=C(CBr)C=C1
Appearance (Color)
Clear colorless to yellow
Refractive index
1.5464 to 1.5484 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text