Bromoacetyl chloride, 95%, Thermo Scientific Chemicals
Bromoacetyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromoacetyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 197990250
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
Add to cart
Bromoacetyl chloride, 95%, Thermo Scientific Chemicals
Catalog number197990250
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
-
Add to cart
Chemical Identifiers
CAS22118-09-8
IUPAC Name2-bromoacetyl chloride
Molecular FormulaC2H2BrClO
InChI KeySYZRZLUNWVNNNV-UHFFFAOYSA-N
SMILESClC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Refractive index1.4940 to 1.4970 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric94.0 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only