Thermo Scientific Chemicals

Potassium carbonate, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 198040250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Potassium carbonate, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 198040250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 198040250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
584-08-7
IUPAC Name
dipotassium carbonate
Molecular Formula
CK2O3
InChI Key
BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]C([O-])=O
Appearance (Form)
Powder or granular powder
Loss on drying
=<1 % (300°C)
Heavy metals (as Pb)
=<5 ppm
Chloride (Cl)
=<30 ppm
Sodium (Na)
=<1000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text