Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Glass bottle
Catalog number 198070010
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
Add to cart
Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Catalog number198070010
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
Particle size=<200 mesh
Appearance (Form)Powder
Arsenic (As)=<100 ppm
Iron (Fe)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only