Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 198072500
Price (USD)/ Each
136.65
Online exclusive
160.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
136.65
Online exclusive
160.00 
Save 23.35 (15%)
Add to cart
Selenium, 99.5+%, powder, -200 mesh, Thermo Scientific Chemicals
Catalog number198072500
Price (USD)/ Each
136.65
Online exclusive
160.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
Molecular Weight (g/mol)78.97
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
Appearance (Form)Powder
Arsenic (As)=<100 ppm
Iron (Fe)=<200 ppm
Lead (Pb)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only