Thermo Scientific Chemicals

Zinc, 98+%, dust (stable acc. to UN classification class 4), Thermo Scientific Chemicals

Catalog number: 198340025
2.5 kg, Each
Thermo Scientific Chemicals

Zinc, 98+%, dust (stable acc. to UN classification class 4), Thermo Scientific Chemicals

Catalog number: 198340025
2.5 kg, Each
Quantity
Catalog number: 198340025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-66-6
IUPAC Name
zinc
Molecular Formula
Zn
InChI Key
HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES
[Zn]
Particle Size/Sieve analysis
=<0.1 % Residue on 250 µm(+60 mesh) sieve
Appearance (Form)
Powder
Zinc (Zn)
>=98.0 %
Appearance (Color)
Grey
Particle Size/Sieve analysis
=<1 % Residue on 150 µm(+100 mesh) sieve

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text