Thermo Scientific Chemicals

Sodium periodate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 198381000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium periodate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 198381000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 198381000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-28-5
IUPAC Name
sodium periodate
Molecular Formula
INaO4
InChI Key
JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES
[Na+].[O-][I](=O)(=O)=O
Appearance (Color)
White to off-white
Titration Iodometric
>=98.5 %
pH
3.4 to 4.5 (5 % at 20°C)
Sulfate (SO4)
=<200 ppm
Iron (Fe)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text