Thermo Scientific Chemicals

Potassium periodate, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 198390025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium periodate, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 198390025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 198390025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-21-8
IUPAC Name
potassium periodate
Molecular Formula
IKO4
InChI Key
FJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES
[K+].[O-][I](=O)(=O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Manganese (Mn)
=<1 ppm
Titration Iodometric
>=99.4 %
Other halogens (as Cl)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text