Periodic acid, 99+%, extra pure
Periodic acid, 99+%, extra pure
Periodic acid, 99+%, extra pure
Periodic acid, 99+%, extra pure
Thermo Scientific Chemicals

Periodic acid, 99+%, extra pure

Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid | CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 198400025
Price (EUR)
1.418,00
Each
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (EUR)
1.418,00
Each
Add to cart
Chemical Identifiers
IUPAC Nametetrahydroxyiodous acid
Molecular FormulaH5IO6
InChI KeyTWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILESO[I](O)(O)(O)(O)=O
Molecular Weight (g/mol)227.94
View more
SpecificationsSpecification SheetSpecification Sheet
IdentificationPositive for periodate
Insoluble matter=<0.01 % (in water)
Sulfate (SO4)=<200 ppm
Appearance (Color)White to light yellow
Titration Iodometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link