Thermo Scientific Chemicals

Aluminum bromide, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 198415000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Aluminum bromide, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 198415000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 198415000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7727-15-3
IUPAC Name
aluminium(3+) tribromide
Molecular Formula
AlBr3
InChI Key
PQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES
[Al+3].[Br-].[Br-].[Br-]
Appearance (Form)
Crystalline powder or chunks
Titration Complexometric
>=98.0 %
Chloride (Cl)
=<2000 ppm
Sulfate (SO4)
=<200 ppm
Appearance (Color)
White to yellow to pink to grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text