2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 198455000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2033-24-1
IUPAC Name2,2-dimethyl-1,3-dioxane-4,6-dione
Molecular FormulaC6H8O4
InChI KeyGXHFUVWIGNLZSC-UHFFFAOYSA-N
SMILESCC1(C)OC(=O)CC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point92°C to 96°C
Water=<0.1 % (K.F.)
Titration with NaOH>=97.5 %
Appearance (Color)White to beige to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only