Thermo Scientific Chemicals

Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 198531000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 198531000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 198531000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10039-54-0
IUPAC Name
bis(hydroxyazanium) sulfate
Molecular Formula
H8N2O6S
InChI Key
VGYYSIDKAKXZEE-UHFFFAOYSA-L
SMILES
[NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %
Appearance (Color)
White
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text