Lead(II) bromide, 98+%, extra pure, Thermo Scientific Chemicals
Lead(II) bromide, 98+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) bromide, 98+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 198851000
Price (USD)/ Each
86.65
Online exclusive
102.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
102.00 
Save 15.35 (15%)
Add to cart
Lead(II) bromide, 98+%, extra pure, Thermo Scientific Chemicals
Catalog number198851000
Price (USD)/ Each
86.65
Online exclusive
102.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameλ²-lead(2+) dibromide
Molecular FormulaBr2Pb
InChI KeyZASWJUOMEGBQCQ-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Pb++]
Molecular Weight (g/mol)367.00
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<100 ppm
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Titration Argentometric>=98.0 %
Loss on drying=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only