Thermo Scientific Chemicals

Lead(II) iodide, 99%, Thermo Scientific Chemicals

Catalog number: 198860500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Lead(II) iodide, 99%, Thermo Scientific Chemicals

Catalog number: 198860500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 198860500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-63-0
IUPAC Name
λ²-lead(2+) diiodide
Molecular Formula
I2Pb
InChI Key
RQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES
[I-].[I-].[Pb++]
Titration Argentometric
>=98.5 %
Chloride (Cl)
=<100 ppm
Appearance (Color)
Yellow to orange
Loss on drying
=<0.1 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text