Zinc chloride, 98.5%, for analysis, Thermo Scientific Chemicals
Zinc chloride, 98.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride, 98.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 198940025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total nitrogen=<0.002 %
Insoluble matter=<0.005 %
Sulfate (SO4)=<20 ppm
Magnesium (Mg)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only