Fusaric acid, 99%, Thermo Scientific Chemicals
Fusaric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fusaric acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 198960010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-butylpyridine-2-carboxylic acid
Molecular FormulaC10H13NO2
InChI KeyDGMPVYSXXIOGJY-UHFFFAOYSA-N
SMILESCCCCC1=CC=C(N=C1)C(O)=O
Molecular Weight (g/mol)179.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Melting point97°C to 101°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only