Thermo Scientific Chemicals

Sulfamide, 98+%, Thermo Scientific Chemicals

Catalog number: 199020050
5 g, Each
Thermo Scientific Chemicals

Sulfamide, 98+%, Thermo Scientific Chemicals

Catalog number: 199020050
5 g, Each
Quantity
Catalog number: 199020050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7803-58-9
IUPAC Name
sulfamoylamine
Molecular Formula
H4N2O2S
InChI Key
NVBFHJWHLNUMCV-UHFFFAOYSA-N
SMILES
NS(N)(=O)=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (50°C, 2 hrs) (vacuum)
Appearance (Color)
White to practically white
Related substances
(TLC) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text