Sulfamide, 98+%, Thermo Scientific Chemicals
Sulfamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 199021000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7803-58-9
IUPAC Namesulfamoylamine
Molecular FormulaH4N2O2S
InChI KeyNVBFHJWHLNUMCV-UHFFFAOYSA-N
SMILESNS(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<0.5 % (50°C, 2 hrs) (vacuum)
Appearance (Color)White to practically white
Related substances(TLC) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only