Thermo Scientific Chemicals

Cerium(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 199125000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cerium(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 199125000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 199125000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1306-38-3
IUPAC Name
dioxocerium
Molecular Formula
CeO2
InChI Key
CETPSERCERDGAM-UHFFFAOYSA-N
SMILES
O=[Ce]=O
Appearance (Color)
White to yellow
Appearance
lanthanum, neodymium, praseodymium, etc.
Appearance
May give a pink to red-brown color due to traces
Appearance (Form)
Powder
Total conc. of imp.as REO
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text