Cerium(IV) sulfate tetrahydrate, 98+%, Thermo Scientific Chemicals
Cerium(IV) sulfate tetrahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(IV) sulfate tetrahydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 199130025
Price (USD)/ Each
2,618.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
3080.00 
Save 461.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
2,618.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
3080.00 
Save 461.35 (15%)
Add to cart
Cerium(IV) sulfate tetrahydrate, 98+%, Thermo Scientific Chemicals
Catalog number199130025
Price (USD)/ Each
2,618.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
3080.00 
Save 461.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10294-42-5
IUPAC Namecerium(3+) tetrahydrate hydrogen sulfate sulfate
Molecular FormulaCeH9O12S2
InChI KeyZBNJQJDKRZKDSV-UHFFFAOYSA-K
SMILESO.O.O.O.[Ce+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<30 ppm
Chloride (Cl)=<100 ppm
Iron (Fe)=<150 ppm
Appearance (Form)Powder
Insoluble matter=<0.1 % (in H2SO4)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only