Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 199160010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 199160010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 199160010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-81-8
IUPAC Name
dilanthanum(3+) trioxidandiide
Molecular Formula
La2O3
InChI Key
MRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[La+3].[La+3]
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Assay
>=99.85 % (La2O3) TREO base
Iron(III)oxide (Fe2O3)
=<250 ppm
Loss on ignition
=<3.0 % (1000°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text