Lanthanum(III) oxide, 99.9%, (trace metal basis)
Lanthanum(III) oxide, 99.9%, (trace metal basis)
Lanthanum(III) oxide, 99.9%, (trace metal basis)
Lanthanum(III) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.9%, (trace metal basis)

CAS: 1312-81-8 | La2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 199162500
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
Assay>=99.85 % (La2O3) TREO base
Iron(III)oxide (Fe2O3)=<250 ppm
Loss on ignition=<3.0 % (1000°C)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472124Certificate of AnalysisMay 17, 2025199162500, 199160010
A0462221Certificate of AnalysisApr 24, 2024199162500, 199160010
A0458171Certificate of AnalysisNov 24, 2023199162500, 199160010
A0452169Certificate of AnalysisMay 02, 2023199162500, 199160010
A0398407Certificate of AnalysisDec 15, 2022199162500, 199160010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link