m-Cresol Purple, sodium salt, pure, water soluble, Thermo Scientific Chemicals
m-Cresol Purple, sodium salt, pure, water soluble, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Cresol Purple, sodium salt, pure, water soluble, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 199250050
Price (USD)/ Each
67.50
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
67.50
Add to cart
m-Cresol Purple, sodium salt, pure, water soluble, Thermo Scientific Chemicals
Catalog number199250050
Price (USD)/ Each
67.50
-
Add to cart
Chemical Identifiers
CAS62625-31-4
IUPAC Namesodium 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular FormulaC21H17NaO5S
InChI KeyALWHNCXHKCVHCK-UHFFFAOYNA-M
SMILES[Na+].CC1=CC(O)=CC=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Visual transition intervalfrom pH 7.4 (yellow) to pH 9.0 (purple)
Appearance (Form)Powder
Solubility(0.05 % in water) Clear
Appearance (Color)Orange-brown to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only