Thermo Scientific Chemicals

Diethylaluminium chloride, 1M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 199521000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Diethylaluminium chloride, 1M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 199521000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 199521000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-10-6
IUPAC Name
chlorodiethylalumane
Molecular Formula
C4H10AlCl
InChI Key
YNLAOSYQHBDIKW-UHFFFAOYSA-M
SMILES
CC[Al](Cl)CC
Appearance (Color)
Colorless
Appearance (Form)
Solution
Strength
0.95 to 1.10 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text