Thermo Scientific Chemicals

Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals

Catalog number: 199570250
25 g, Each
Thermo Scientific Chemicals

Rhodium on alumina, 5% Rh, powder, Thermo Scientific Chemicals

Catalog number: 199570250
25 g, Each
Quantity
Catalog number: 199570250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-16-6
IUPAC Name
rhodium
Molecular Formula
Rh
InChI Key
MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES
[Rh]
Appearance (Form)
Powder
Assay
5 % (Rh) Typical
Appearance (Color)
Brown or grey to black

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text