Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 199592500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3153-26-2
IUPAC Nameoxovanadiumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O5V
InChI KeyJFHJZWAQYMGNBE-SUKNRPLKSA-L
SMILESO=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay>=98.5 %
Appearance (Color)Green to blue-green
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only