Thermo Scientific Chemicals

Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals

Catalog number: 199620100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals

Catalog number: 199620100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 199620100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12135-22-7
IUPAC Name
palladium(2+) dihydroxide
Molecular Formula
H2O2Pd
InChI Key
NXJCBFBQEVOTOW-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Pd++]
Appearance (Color)
Black
Water
45.0 to 55.0 %
Palladium (Pd)
18.0 to 22.0 % On dry basis
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text