Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals
Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 199620500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12135-22-7
IUPAC Namepalladium(2+) dihydroxide
Molecular FormulaH2O2Pd
InChI KeyNXJCBFBQEVOTOW-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Water45.0 to 55.0 %
Palladium (Pd)18.0 to 22.0 % On dry basis
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only