Thermo Scientific Chemicals

Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals

Catalog number: 199645000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals

Catalog number: 199645000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 199645000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-89-6
IUPAC Name
λ¹-copper(1+) chloride
Molecular Formula
ClCu
InChI Key
OXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES
[Cl-].[Cu+]
Appearance (Color)
Green-grey
Appearance (Form)
Powder or pearls
Iron (Fe)
=<400 ppm
Sulfate (SO4)
=<500 ppm
Titration Complexometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text