Thermo Scientific Chemicals

Benzeneseleninic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 199710250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzeneseleninic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 199710250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 199710250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17697-12-0
IUPAC Name
benzeneseleninyl benzeneseleninate
Molecular Formula
C12H10O3Se2
InChI Key
FHPZOWOEILXXBD-UHFFFAOYSA-N
SMILES
O=[Se](O[Se](=O)C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or needles
Titration with NaOH
>=97.5 %
Appearance (Color)
White to yellow to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text