Thermo Scientific Chemicals

Citric acid, trilithium salt tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 199855000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Citric acid, trilithium salt tetrahydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 199855000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 199855000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6080-58-6
IUPAC Name
trilithium(1+) 2-hydroxypropane-1,2,3-tricarboxylate tetrahydrate
Molecular Formula
C6H13Li3O11
InChI Key
HXGWMCJZLNWEBC-UHFFFAOYSA-K
SMILES
[Li+].[Li+].[Li+].O.O.O.O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Form)
Crystalline powder
Titration with HClO4
>=97.5 % (On anhydrous substance)
Appearance (Color)
White
Infrared spectrum
Conforms
Water
=<24 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text