Thermo Scientific Chemicals

Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 199870100
10 kg, Each
Thermo Scientific Chemicals

Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 199870100
10 kg, Each
Quantity
Catalog number: 199870100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-35-8
IUPAC Name
lithium(1+) bromide
Molecular Formula
BrLi
InChI Key
AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES
[Li+].[Br-]
Titration Argentometric
>=99.0 %
Sulfate (SO4)
=<200 ppm
Copper (Cu)
=<3 ppm
Magnesium (Mg)
=<20 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text