Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals

Lithium bromide, 99+%, for analysis, anhydrous

CAS: 7550-35-8 | BrLi | 86.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
10 kg
Catalog number 199875000
Price (USD)
159.65
Special offer
Online exclusive
Ends: 26-Sep-2025
197.00
Save 37.35 (19%)
Each
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
159.65
Special offer
Online exclusive
Ends: 26-Sep-2025
197.00
Save 37.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Sulfate (SO4)=<200 ppm
Copper (Cu)=<3 ppm
Magnesium (Mg)=<20 ppm
Appearance (Color)White
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link