Thermo Scientific Chemicals

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Catalog number: 199880100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Catalog number: 199880100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 199880100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7447-41-8
IUPAC Name
lithium(1+) chloride
Molecular Formula
ClLi
InChI Key
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES
[Li+].[Cl-]
Appearance (Form)
Crystalline powder or crystals or granules
Sulfate (SO4)
=<0.01 %
Potassium (K)
=<0.0015 %
Appearance (Color)
White
Titration Argentometric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text