Sulfur, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Sulfur, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number 199930500
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
Add to cart
Sulfur, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Catalog number199930500
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pieces
Particle size=<12 mm
Total trace metal impurities=<0.001 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only