Thermo Scientific Chemicals

Calcium hydride, ca. 93%, extra pure, 10-100 mm pieces, up to 10% powder, Thermo Scientific Chemicals

Catalog number: 199961000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium hydride, ca. 93%, extra pure, 10-100 mm pieces, up to 10% powder, Thermo Scientific Chemicals

Catalog number: 199961000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 199961000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-78-8
IUPAC Name
calcium
Molecular Formula
Ca
InChI Key
OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES
[Ca]
Appearance (Color)
Grey
Magnesium (Mg)
=<8000 ppm
Aluminium (Al)
=<100 ppm
Hydrogen (H)
>=980 ml/g CaH2
Particle size
10 to 100 mm (containing up to 10% fines)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text