Ammonium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Ammonium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium bromide, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 199982500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 % (on dry substance)
Sulfate (SO4)=<0.005 %
pH4.5 to 6.0 (5 % solution)
Iron (Fe)=<1 ppm
Nickel (Ni)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only