Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Have Questions?
Catalog number 1R120434-0010
Price (USD)/ Each
-
Request A Quote
Description:
Spring Loaded Transfer Nut
For Use With (Equipment):
Thermo Scientific ISQ and TSQ
Specifications
DescriptionSpring Loaded Transfer Nut
For Use With (Equipment)Thermo Scientific ISQ and TSQ