Search Thermo Fisher Scientific
Helping laboratories achieve reliable and high-throughput GC-MS analysis of pesticides, the automated cartridge-based micro Solid Phase Extraction (μSPE) Clean-up Tool for QuEChERS extracts is available for the TriPlus™ RSH and TriPlus RSH SMART Autosamplers. This tool offers a fully automated on-line approach for clean-up of QuEChERS extracts, and replaces the dispersive SPE manual procedure. Suitable for a wide variety of food matrices, even those with high lipid contents, the μSPE tool offers a unified method for GC-MS analysis of hundreds of pesticides and organic contaminants. By using a single type of miniaturized cartridge, this tool also offers the benefits of scaling down sample volume and solvent usage.
Enhancing the automated workflow capability of the TriPlus RSH and TriPlus RSH SMART autosamplers, the μSPE Clean-up Tool for QuEChERS extracts provides today's analytical testing laboratories with the productivity they need to succeed. Unlike classic vacuum manifold-based SPE, the flow rates applied to the μSPE cartridge can be precisely controlled with a liquid syringe by the autosampler, assuring optimum recovery. Low-flow rates of ∼2 μL/s in the load and elution steps are used for sharp analyte/matrix separation.
Other advantages of the μSPE Clean-up Tool include the following: