Search Thermo Fisher Scientific
Easily create and test sample preparation methods with the Thermo Scientific™ TriPlus™ RSH Sampling Workflow Editor Software. When connected with the Thermo Scientific™ TriPlus™ RSH and TriPlus RSH SMART, this software enables easy programming of the autosampler to fully automate common sample handling operations as well as more complex sample preparation procedures. The visual programming approach of the software makes this operation very easy and intuitive via its drag and drop functionality, not requiring any programming skills.
Type | Chromatography Software |
For Use With (Equipment) | TriPlus RSH and TriPlus RSH SMART Autosamplers |
Description | TriPlus RSH Sampling Workflow Editor Software Tool |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Type | For Use With (Equipment) | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1R77010-1200 | Each | TriPlus RSH Sampling Workflow Editor Software Tool | Chromatography Software | TriPlus RSH and TriPlus RSH SMART Autosamplers | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Simply drag-and-drop to program your own sample preparation method, and let the TriPlus RSH Sampling Workflow Editor Software do the rest. This tool's many benefits include:
Intuitive Programming