2.5EVERGREEN Full specifications
Each 2.5 oz. (74mL); Evergreen Standard Polypropylene Container Request A Quote - - Description 2.5 oz. (74mL); Evergreen Diameter (Metric) Inner 50 mm Color Evergreen Material Polypropylene Sterility Non-sterile Volume (Metric) 74 mL No. per Case 400 Style Standard Volume (English) 2.5 oz. Closure Type Flip Top Height (Metric) 50 mm Type Polypropylene Container Unit Size Each
0385 Full specifications
Case of 400 90mL (3 oz.); With skirt top Skirt top Polypropylene Container Request A Quote - - Description 90mL (3 oz.); With skirt top Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Skirt top Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
03CLFREGORED Full specifications
Case of 400 3 oz. (90mL); Frego Red Standard Polypropylene Container Request A Quote - - Description 3 oz. (90mL); Frego Red Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Standard Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 400
03EDM33 Full specifications
Case of 400 90mL (3 oz.); With write-on space Write-on space Polypropylene Container - Description 90mL (3 oz.); With write-on space Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Write-on space Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
04HP-SMOOTH Full specifications
Case of 250 4 oz. (118mL); High profile; With write-on space High profile with write-on space Polypropylene Container Contact Us
- - Description 4 oz. (118mL); High profile; With write-on space Diameter (Metric) Inner 43 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style High profile with write-on space Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 90 mm Type Polypropylene Container Unit Size Case of 250
04LP Full specifications
Case of 250 4 oz. (118mL); Low profile; Natural Low profile Polypropylene Container - Description 4 oz. (118mL); Low profile; Natural Diameter (Metric) Inner 63 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style Low profile Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 50 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 250
05WIDE Full specifications
Case of 175 5 oz. (148mL); Wide mouth; Natural Low profile, wide mouth Polypropylene Container - Description 5 oz. (148mL); Wide mouth; Natural Diameter (Metric) Inner 62 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 148 mL No. per Case 175 Style Low profile, wide mouth Volume (English) 5 oz. Closure Type Flip Top Height (Metric) 70 mm Type Polypropylene Container Unit Size Case of 175
08CL Full specifications
Case of 150 8 oz. (236mL); Natural Standard Polypropylene Container - Description 8 oz. (236mL); Natural Diameter (Metric) Inner 75 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 236 mL No. per Case 150 Style Standard Volume (English) 8 oz. Closure Type Flip Top Height (Metric) 75 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 150
2.5CL Full specifications
Case of 400 2.5 oz. (74mL); Natural Standard Polypropylene Container - Description 2.5 oz. (74mL); Natural Diameter (Metric) Inner 50 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 74 mL No. per Case 400 Style Standard Volume (English) 2.5 oz. Closure Type Flip Top Height (Metric) 50 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 400
2.5RED Full specifications
Each 2.5 oz. (74mL); Red Standard Polypropylene Container Contact Us
- - Description 2.5 oz. (74mL); Red Diameter (Metric) Inner 50 mm Color Evergreen Material Polypropylene Sterility Non-sterile Volume (Metric) 74 mL No. per Case 400 Style Standard Volume (English) 2.5 oz. Closure Type Flip Top Height (Metric) 50 mm Type Polypropylene Container Unit Size Each